Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENOPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292017.1 | NP_001278946.1 | 4 | 765 | ||
NM_021204.4 | NP_067027.1 | 4 | 794 | |||
NR_120457.1 | 3 | 562 | ||||
GenBank mRNA | AF087881.1 | - | 4 | 664 | ||
AF113125.1 | - | 4 | 665 | |||
AF177286.1 | - | 2 | 231 | |||
AK022656.1 | - | 4 | 660 | |||
BC001317.1 | - | 3 | 380 | |||
BC065815.1 | - | 4 | 632 | 6526329 | ||
CN335976.1 | - | 4 | 606 | |||
CR457141.1 | - | 4 | 444 |