Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGAP22 SIRNA Assays ›
Gene Symbol:
ARHGAP22
Gene Name:
Rho GTPase activating protein 22
Gene Aliases:
RhoGAP2, RhoGap22
Chromosome Location:
Chr.10: 48429831 - 48656265 on Build GRCh38
UniGene ID:
Hs.655672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33880
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGAP22
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256024.1
NP_001242953.1
8
1274
NM_001256025.2
NP_001242954.1
8
1140
NM_001256026.1
NP_001242955.1
6
780
NM_021226.3
NP_067049.2
8
1226
NR_045675.1
9
1725
XM_005270014.3
XP_005270071.1
6
684
XM_011540002.1
XP_011538304.1
9
1825
XM_011540003.1
XP_011538305.1
9
1127
XM_011540004.1
XP_011538306.1
9
1777
XM_011540005.1
XP_011538307.1
6
828
XM_011540006.1
XP_011538308.1
8
1648
XM_011540007.2
XP_011538309.1
6
5032
XM_011540011.2
XP_011538313.1
5
507
XM_011540012.2
XP_011538314.1
6
1185
XM_011540013.2
XP_011538315.1
9
1825
XM_011540015.2
XP_011538317.1
9
1825
XM_017016470.1
XP_016871959.1
7
1094
XM_017016471.1
XP_016871960.1
6
1099
XM_017016472.1
XP_016871961.1
6
672
XM_017016473.1
XP_016871962.1
9
1777
XM_017016474.1
XP_016871963.1
8
1223
XM_017016475.1
XP_016871964.1
7
1094
XM_017016476.1
XP_016871965.1
8
1223
XM_017016477.1
XP_016871966.1
8
1171
GenBank mRNA
AK055037.1
-
7
819
AK055937.1
-
9
1725
AK127586.1
-
6
780
AK293579.1
-
8
1026
AY324801.1
-
8
1188
BC047096.1
-
8
1226
BC126444.1
-
8
1095
8992160
BC136319.1
-
8
1095
9020306
EF560749.1
-
8
1198
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
cell differentiation
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0