Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021249.3 | NP_067072.3 | 11 | 1101 | ||
NM_152233.2 | NP_689419.2 | 12 | 996 | |||
XM_006720224.1 | XP_006720287.1 | 11 | 883 | |||
GenBank mRNA | AF109364.1 | - | 7 | 479 | ||
AF121856.1 | - | 12 | 987 | |||
AK021456.1 | - | 7 | 483 | |||
AK292250.1 | - | 12 | 979 | |||
AK296213.1 | - | 13 | 1124 | |||
AK308464.1 | - | 13 | 1124 | |||
BC001798.1 | - | 11 | 1101 | 3354467 | ||
BI870378.1 | - | 6 | 377 | |||
U83194.1 | - | 12 | 964 |