Search
Search

GALNT11| Gene Symbol: GALNT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304514.1 | NP_001291443.1 | 8 | 1177 | ||
| NM_022087.3 | NP_071370.2 | 9 | 1510 | |||
| XM_006716082.2 | XP_006716145.1 | 9 | 1421 | |||
| XM_006716083.2 | XP_006716146.1 | 9 | 1389 | |||
| XM_006716084.2 | XP_006716147.1 | 9 | 1353 | |||
| XM_017012500.1 | XP_016867989.1 | 8 | 1367 | |||
| GenBank mRNA | AK025287.1 | - | 9 | 1344 | ||
| AK026056.1 | - | 5 | 583 | |||
| AK124934.1 | - | 11 | 1698 | |||
| AK128545.1 | - | 4 | 1336 | |||
| AK298911.1 | - | 8 | 1100 | |||
| Y12434.3 | - | 8 | 1261 | |||