siRNA Details
Gene Symbol: SOX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022454.3 | NP_071899.1 | 2 | 2231 |

Gene Symbol: SOX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022454.3 | NP_071899.1 | 2 | 2231 |