Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022465.3 | NP_071910.3 | 2 | 735 | ||
XM_005269086.3 | XP_005269143.1 | 4 | 277 | |||
XM_011538664.1 | XP_011536966.1 | 4 | 280 | |||
XM_011538666.2 | XP_011536968.1 | 3 | 128 | |||
XM_017019805.1 | XP_016875294.1 | 6 | 525 | |||
XM_017019806.1 | XP_016875295.1 | 6 | 768 | |||
XM_017019807.1 | XP_016875296.1 | 6 | 765 | |||
XM_017019808.1 | XP_016875297.1 | 5 | 450 | |||
XM_017019809.1 | XP_016875298.1 | 6 | 522 | |||
XM_017019810.1 | XP_016875299.1 | 6 | 620 | |||
XM_017019811.1 | XP_016875300.1 | 5 | 447 | |||
XM_017019812.1 | XP_016875301.1 | 4 | 523 | |||
XM_017019813.1 | XP_016875302.1 | 4 | 520 | |||
GenBank mRNA | BX647761.1 | - | 3 | 305 | ||
DA797441.1 | - | 2 | 299 |