siRNA Details
Gene Symbol: ZMAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022470.3 | NP_071915.1 | 5 | 943 | ||
NM_152240.2 | NP_689426.1 | 6 | 1064 | |||
XM_005247706.2 | XP_005247763.1 | 5 | 1354 | |||
XM_011513073.1 | XP_011511375.1 | 6 | 1475 | |||
GenBank mRNA | AF355465.1 | - | 5 | 752 | ||
AK022358.1 | - | 5 | 870 | |||
AK122768.1 | - | 6 | 953 | |||
AY037945.1 | - | 4 | 642 | |||
BC002896.2 | - | 5 | 871 | 3943641 |

More Information
Panther Classification:
Molecular Function -




