Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DCLRE1C SIRNA Assays ›
          Gene Symbol:
          DCLRE1C
          Gene Name:
          DNA cross-link repair 1C
          Gene Aliases:
          A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C
          Chromosome Location:
          Chr.10: 14897853 - 14954432 on Build GRCh38
          UniGene ID:
          Hs.655932
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s34728

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DCLRE1C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001033855.2 NP_001029027.1 14 2285
          NM_001033857.2 NP_001029029.1 15 2370
          NM_001033858.2 NP_001029030.1 16 2692
          NM_001289076.1 NP_001276005.1 12 2144
          NM_001289077.1 NP_001276006.1 15 2416
          NM_001289078.1 NP_001276007.1 12 2177
          NM_001289079.1 NP_001276008.1 16 2738
          NM_022487.3 NP_071932.2 13 2229
          NR_110297.1 17 2638
          XM_006717491.3 XP_006717554.1 14 1959
          XM_011519616.1 XP_011517918.1 14 1975
          XM_011519617.1 XP_011517919.1 15 2094
          XM_011519619.1 XP_011517921.1 16 2121
          XM_017016557.1 XP_016872046.1 14 1987
          XM_017016558.1 XP_016872047.1 15 2033
          GenBank mRNA AF088055.1 - 1 524
          AF395747.1 - 15 1986
          AF395748.1 - 15 2032
          AF395749.1 - 13 1845
          AF395750.1 - 12 1793
          AF395751.1 - 16 2308
          AF395752.1 - 16 2354
          AJ296101.1 - 14 1901
          AK021422.1 - 13 1892
          AK022922.1 - 16 2291
          AK093757.1 - 3 1498
          AK290598.1 - 14 1948
          BC000863.1 - 1 665
          BC009185.2 - 12 1768 3945623
          BX648858.1 - 15 2002

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          double-strand break repair via nonhomologous end joining
          response to ionizing radiation
          B cell differentiation
          protection from non-homologous end joining at telomere
          V(D)J recombination
          interstrand cross-link repair
          nucleic acid phosphodiester bond hydrolysis
          single-stranded DNA endodeoxyribonuclease activity
          damaged DNA binding
          endonuclease activity
          protein binding
          5'-3' exonuclease activity
          5'-3' exodeoxyribonuclease activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0