siRNA Details
Gene Symbol: CRTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042574.2 | NP_001036039.1 | 15 | 1852 | ||
NM_022769.4 | NP_073606.3 | 15 | 1855 | |||
XM_005254968.2 | XP_005255025.1 | 15 | 1823 | |||
XM_011521906.2 | XP_011520208.1 | 14 | 1727 | |||
GenBank mRNA | AK025521.1 | - | 8 | 1019 | ||
AK074170.1 | - | 12 | 1323 | |||
AK090443.1 | - | 15 | 1812 | |||
AK225300.1 | - | 8 | 1019 | |||
AY360173.1 | - | 15 | 1676 | |||
BC074730.2 | - | 6 | 795 | |||
BC074731.2 | - | 6 | 798 | 30915456 | ||
CN298837.1 | - | 3 | 170 |

More Information
Panther Classification:
Molecular Function -



