Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDRG4 SIRNA Assays ›
Gene Symbol:
NDRG4
Gene Name:
NDRG family member 4
Gene Aliases:
BDM1, SMAP-8, SMAP8
Chromosome Location:
Chr.16: 58463645 - 58515403 on Build GRCh38
UniGene ID:
Hs.322430
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35157
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDRG4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130487.1
NP_001123959.1
4, 5
623
NM_001242833.1
NP_001229762.1
3, 4
322
NM_001242834.1
NP_001229763.1
2, 3
286
NM_001242835.1
NP_001229764.1
2, 3
232
NM_001242836.1
NP_001229765.1
2, 3
232
NM_020465.3
NP_065198.1
4, 5
398
NM_022910.3
NP_075061.1
4, 5
495
NR_040072.1
2, 3
324
XM_006721252.3
XP_006721315.1
5, 6
498
XM_006721253.3
XP_006721316.1
5, 6
431
XM_006721254.3
XP_006721317.1
4, 5
485
XM_006721256.3
XP_006721319.1
5, 6
429
XM_006721257.3
XP_006721320.1
4, 5
470
XM_006721258.3
XP_006721321.1
4, 5
374
XM_006721259.3
XP_006721322.1
4, 5
439
XM_006721260.3
XP_006721323.1
4, 5
377
XM_006721261.3
XP_006721324.1
3, 4
322
XM_006721262.3
XP_006721325.1
2, 3
403
XM_011523290.2
XP_011521592.1
5, 6
585
XM_011523291.2
XP_011521593.1
4, 5
549
XM_011523292.2
XP_011521594.1
5, 6
585
XM_011523293.2
XP_011521595.1
4, 5
543
XM_011523294.2
XP_011521596.1
5, 6
364
XM_017023582.1
XP_016879071.1
4, 5
549
XM_017023583.1
XP_016879072.1
5, 6
585
XM_017023584.1
XP_016879073.1
5, 6
498
XM_017023585.1
XP_016879074.1
4, 5
485
XM_017023586.1
XP_016879075.1
4, 5
432
XM_017023587.1
XP_016879076.1
5, 6
429
XM_017023588.1
XP_016879077.1
4, 5
374
XM_017023589.1
XP_016879078.1
4, 5
439
XM_017023590.1
XP_016879079.1
5, 6
364
GenBank mRNA
AB021172.1
-
4, 5
298
AB033006.2
-
2, 3
218
AB044944.1
-
2, 3
232
AB044945.1
-
2, 3
232
AB044946.1
-
3, 4
245
AF308608.1
-
2, 3
239
AK055038.1
-
2, 3
234
AK126574.1
-
4, 5
395
AK126729.1
-
5, 6
2345
AK296415.1
-
2, 3
286
AK299949.1
-
3, 4
322
AK316089.1
-
2, 3
324
AL136584.1
-
2, 3
218
BC001527.1
-
2, 3
216
BC011795.2
-
2, 3
220
4139594
CF552009.1
-
4, 5
341
CR936733.1
-
4, 5
309
DA069318.1
-
4, 5
456
DA242460.1
-
4, 5
398
KU178760.1
-
2, 3
126
KU178761.1
-
2, 3
126
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
heart looping
response to stress
brain development
visual learning
negative regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of neuron projection development
negative regulation of smooth muscle cell migration
cell growth
cell differentiation
embryonic heart tube development
vesicle docking
negative regulation of smooth muscle cell proliferation
cardiac muscle cell proliferation
cell migration involved in heart development
positive regulation of ERK1 and ERK2 cascade
regulation of endocytic recycling
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0