Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 34 | 5818 | ||
XM_005246787.3 | XP_005246844.1 | 34 | 5844 | |||
XM_005246788.2 | XP_005246845.1 | 13 | 2320 | |||
XM_006712698.3 | XP_006712761.1 | 35 | 5931 | |||
XM_006712699.3 | XP_006712762.1 | 35 | 5931 | |||
XM_011511658.2 | XP_011509960.1 | 35 | 5886 | |||
XM_011511659.2 | XP_011509961.1 | 35 | 5943 | |||
XM_011511660.2 | XP_011509962.1 | 31 | 5265 | |||
XM_011511662.2 | XP_011509964.1 | 27 | 4600 | |||
XM_011511663.2 | XP_011509965.1 | 35 | 5921 | |||
XM_017004728.1 | XP_016860217.1 | 22 | 3998 | |||
XM_017004729.1 | XP_016860218.1 | 10 | 2012 | |||
GenBank mRNA | AK127772.1 | - | 8 | 1686 | ||
AK128636.1 | - | 4 | 545 | |||
AY172970.1 | - | 4 | 724 |