Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324475.1 | NP_001311404.1 | 16 | 1810 | ||
NM_023015.4 | NP_075391.3 | 15 | 2151 | |||
XM_005245459.2 | XP_005245516.1 | 15 | 2168 | |||
XM_005245461.2 | XP_005245518.1 | 9 | 841 | |||
XM_006711490.3 | XP_006711553.1 | 16 | 1621 | |||
XM_011509906.2 | XP_011508208.1 | 15 | 1533 | |||
XM_017002132.1 | XP_016857621.1 | 5 | 409 | |||
GenBank mRNA | AK074979.1 | - | 8 | 849 | ||
AK091908.1 | - | 2 | 2072 | |||
AK290025.1 | - | 16 | 1804 | |||
AK298746.1 | - | 3 | 301 | |||
AK304874.1 | - | 9 | 1240 | |||
AL080172.1 | - | 10 | 1050 | |||
AL832133.1 | - | 14 | 3186 | |||
BC054513.1 | - | 2 | 187 | |||
BC098431.1 | - | 11 | ||||
BC105092.1 | - | 15 | 1614 | 8144095 | ||
BC105094.1 | - | 15 | 1614 | 8144097 | ||
BC116458.1 | - | 15 | 2146 | 5752962 | ||
BX640786.1 | - | 15 | 2137 | |||
BX640950.1 | - | 15 | 2152 | |||
CR749212.1 | - | 13 | 3678 | |||
CR749376.1 | - | 14 | 3803 |