Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127457.2 | NP_001120929.1 | 3 | 298 | ||
NM_021117.3 | NP_066940.2 | 3 | 434 | |||
GenBank mRNA | AB014558.1 | - | 3 | 338 | ||
AK291260.1 | - | 3 | 365 | |||
AK294904.1 | - | 3 | 292 | |||
AK302865.1 | - | 3 | 434 | |||
BC035161.1 | - | 5 | 697 | 5265444 | ||
BC041814.1 | - | 3 | 362 | 5266781 | ||
DC330290.1 | - | 3 | 298 | |||
EU219622.1 | - | 3 | 357 |