Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024040.2 | NP_076945.2 | 5 | 453 | ||
GenBank mRNA | AB025425.1 | - | 4 | 302 | ||
AB232358.1 | - | 4 | 302 | |||
BC000262.1 | - | 5 | 383 | 3353174 | ||
BM756951.1 | - | 5 | 453 | |||
CX165866.1 | - | 5 | 337 |