Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPSCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251888.1 | NP_001238817.1 | 9 | 1235 | ||
NM_024083.3 | NP_076988.1 | 9 | 1235 | |||
NR_045351.1 | 8 | 1179 | ||||
XM_011523601.2 | XP_011521903.1 | 9 | 1235 | |||
XM_011523602.2 | XP_011521904.1 | 9 | 1235 | |||
XM_017025036.1 | XP_016880525.1 | 9 | 1235 | |||
XM_017025037.1 | XP_016880526.1 | 9 | 1235 | |||
XM_017025038.1 | XP_016880527.1 | 10 | 1246 | |||
XM_017025039.1 | XP_016880528.1 | 10 | 1163 | |||
XM_017025040.1 | XP_016880529.1 | 9 | 1100 | |||
XM_017025041.1 | XP_016880530.1 | 8 | 1103 | |||
XM_017025042.1 | XP_016880531.1 | 9 | 1235 | |||
XM_017025043.1 | XP_016880532.1 | 8 | 1103 | |||
XM_017025044.1 | XP_016880533.1 | 10 | 1246 | |||
GenBank mRNA | AF324219.2 | - | 9 | 1160 | ||
AK057403.1 | - | 9 | 1235 | |||
AK057851.1 | - | 8 | 1087 | |||
AK090682.1 | - | 2 | 2600 | |||
AK126967.1 | - | 2 | 4895 | |||
AK127159.1 | - | 2 | 1807 | |||
AK226086.1 | - | 9 | 1108 | |||
AK290624.1 | - | 9 | 1151 | |||
BC006152.1 | - | 9 | 1190 | 3357489 | ||
BC018722.1 | - | 9 | 1140 | 4640076 |