Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024097.3 | NP_077002.2 | 4 | 393 | ||
NR_040733.1 | 3 | 277 | ||||
GenBank mRNA | BC001508.2 | - | 4 | 369 | 2959109 | |
BC001711.1 | - | 4 | 369 | 3543543 | ||
BM702424.1 | - | 3 | 229 | |||
CN256666.1 | - | 3 | 177 |