Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC25A23 SIRNA Assays ›
Gene Symbol:
SLC25A23
Gene Name:
solute carrier family 25 member 23
Gene Aliases:
APC2, MCSC2, SCaMC-3
Chromosome Location:
Chr.19: 6436081 - 6460221 on Build GRCh38
UniGene ID:
Hs.356231
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35549
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC25A23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024103.2
NP_077008.2
3, 4
505
XM_011528274.1
XP_011526576.1
3, 4
956
XM_011528275.1
XP_011526577.1
3, 4
956
XM_011528276.1
XP_011526578.1
3, 4
956
XM_011528277.1
XP_011526579.1
3, 4
956
XM_011528278.1
XP_011526580.1
3, 4
956
XM_011528279.1
XP_011526581.1
3, 4
956
XM_011528280.1
XP_011526582.1
3, 4
956
XM_011528281.1
XP_011526583.1
3, 4
956
XM_011528282.1
XP_011526584.1
3, 4
956
XM_011528283.1
XP_011526585.1
3, 4
956
XM_011528284.1
XP_011526586.1
3, 4
956
XM_011528285.1
XP_011526587.1
3, 4
956
XM_017027285.1
XP_016882774.1
3, 4
956
XM_017027286.1
XP_016882775.1
3, 4
956
XM_017027287.1
XP_016882776.1
3, 4
956
XM_017027288.1
XP_016882777.1
3, 4
956
XM_017027289.1
XP_016882778.1
3, 4
956
XM_017027290.1
XP_016882779.1
3, 4
956
XM_017027291.1
XP_016882780.1
3, 4
956
XM_017027292.1
XP_016882781.1
3, 4
956
GenBank mRNA
AJ512835.1
-
3, 4
470
AJ619962.1
-
3, 4
460
AJ619988.1
-
3, 4
470
AK054901.1
-
2, 3
202
AK294514.1
-
3, 4
363
AY750170.1
-
3, 4
470
BC001656.1
-
3, 4
486
3356718
BG706035.1
-
3, 4
532
Back To Top
More Information
Panther Classification:
Molecular Function -
mitochondrial carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of oxidative phosphorylation
translation
mitochondrial calcium ion transport
calcium ion transmembrane import into mitochondrion
regulation of cellular respiration
regulation of sequestering of calcium ion
adenine nucleotide transport
cellular response to calcium ion
urea homeostasis
structural constituent of ribosome
calcium ion binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0