Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC28B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301011.1 | NP_001287940.1 | 4, 5 | 868 | ||
NM_024296.4 | NP_077272.2 | 4, 5 | 868 | |||
XM_011542113.2 | XP_011540415.1 | 4, 5 | 1627 | |||
XM_017002307.1 | XP_016857796.1 | 4, 5 | 1622 | |||
GenBank mRNA | AK225113.1 | - | 4, 5 | 638 | ||
AK291904.1 | - | 4, 5 | 878 | |||
BC022848.2 | - | 4, 5 | 602 | 3946778 |