Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005210.3 | NP_005201.2 | 2 | 88 | ||
XM_017003402.1 | XP_016858891.1 | 2 | 262 | |||
GenBank mRNA | BC074944.2 | - | 2 | 88 | 30915273 | |
BC074945.2 | - | 2 | 88 | 30915496 | ||
BC117384.1 | - | 2 | 88 | 40125935 | ||
BC117388.1 | - | 2 | 88 | 40125939 | ||
BY794984.2 | - | 2 | 103 | |||
KT326704.1 | - | 2 | 93 |