Search
Search

PGBD5| Gene Symbol: PGBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258311.1 | NP_001245240.1 | 7 | 1393 | ||
| XM_017002330.1 | XP_016857819.1 | 7 | 2462 | |||
| GenBank mRNA | AK021475.1 | - | 3 | 148 | ||
| AK297321.1 | - | 4 | 362 | |||
| AK315968.1 | - | 4 | 423 | |||
| AL833243.1 | - | 6 | 1038 | |||
| BC013901.2 | - | 3 | 202 | |||
| BC027466.1 | - | 4 | 362 | |||
| BC036865.1 | - | 6 | 820 | |||
| BC150638.1 | - | 7 | 1210 | 9057008 | ||
| KF670820.1 | - | 7 | 1194 | |||