Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP7D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173516.1 | NP_001166987.1 | 16, 17 | 2589 | ||
NM_001173517.1 | NP_001166988.1 | 15, 16 | 2596 | |||
NM_024597.3 | NP_078873.2 | 16, 17 | 2701 | |||
XM_005262472.1 | XP_005262529.1 | 16, 17 | 2553 | |||
XM_017029843.1 | XP_016885332.1 | 16, 17 | 2553 | |||
GenBank mRNA | AK022711.1 | - | 4, 5 | 382 | ||
AK301478.1 | - | 16, 17 | 2590 | |||
AL832120.1 | - | 15, 16 | 2388 | |||
AL832475.1 | - | 6, 7 | 704 | |||
BC040518.1 | - | 16, 17 | 2565 | |||
BC064350.1 | - | 16, 17 | 2381 |