Search
Search

CSF2RB| Gene Symbol: CSF2RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000395.2 | NP_000386.1 | 9 | 1256 | ||
| XM_005261340.3 | XP_005261397.1 | 9 | 1333 | |||
| XM_011529903.2 | XP_011528205.1 | 9 | 1333 | |||
| XM_011529904.2 | XP_011528206.1 | 9 | 1315 | |||
| XM_011529905.2 | XP_011528207.1 | 9 | 1333 | |||
| GenBank mRNA | AK302993.1 | - | 7 | 918 | ||
| BC070085.1 | - | 9 | 1256 | 30344148 | ||
| CR456428.1 | - | 8 | 1130 | |||
| M59941.1 | - | 8 | 1067 | |||