Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36158
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
5
569
NM_001164629.3
NP_001158101.1
5
554
NM_001284233.1
NP_001271162.1
3
378
NM_001284234.1
NP_001271163.1
3
269
NM_001284235.1
NP_001271164.1
5
536
NM_001284238.1
NP_001271167.1
7
768
NM_024659.4
NP_078935.2
5
536
XM_005263774.2
XP_005263831.1
5
506
XM_005263782.2
XP_005263839.1
5
522
XM_005263784.2
XP_005263841.1
3
381
XM_011511843.2
XP_011510145.1
5
555
XM_011511855.2
XP_011510157.1
3
499
XM_011511856.2
XP_011510158.1
3
281
XM_017004917.1
XP_016860406.1
9
1177
XM_017004918.1
XP_016860407.1
7
769
XM_017004919.1
XP_016860408.1
5
655
XM_017004920.1
XP_016860409.1
7
3811
XM_017004921.1
XP_016860410.1
6
604
XM_017004922.1
XP_016860411.1
6
781
XM_017004923.1
XP_016860412.1
5
569
XM_017004924.1
XP_016860413.1
7
765
XM_017004925.1
XP_016860414.1
4
427
XM_017004926.1
XP_016860415.1
3
505
XM_017004927.1
XP_016860416.1
5
536
XM_017004928.1
XP_016860417.1
7
769
XM_017004929.1
XP_016860418.1
5
506
XM_017004930.1
XP_016860419.1
5
1157
XM_017004931.1
XP_016860420.1
5
570
XM_017004932.1
XP_016860421.1
5
540
XM_017004933.1
XP_016860422.1
5
522
XM_017004934.1
XP_016860423.1
6
3729
XM_017004935.1
XP_016860424.1
7
1599
XM_017004936.1
XP_016860425.1
6
776
XM_017004937.1
XP_016860426.1
3
510
XM_017004938.1
XP_016860427.1
3
508
XM_017004939.1
XP_016860428.1
5
558
XM_017004940.1
XP_016860429.1
3
506
XM_017004941.1
XP_016860430.1
3
505
XM_017004942.1
XP_016860431.1
5
552
XM_017004943.1
XP_016860432.1
5
1218
XM_017004944.1
XP_016860433.1
5
551
XM_017004945.1
XP_016860434.1
7
769
XM_017004946.1
XP_016860435.1
5
506
XM_017004947.1
XP_016860436.1
5
1189
XM_017004948.1
XP_016860437.1
6
3729
XM_017004949.1
XP_016860438.1
3
504
XM_017004950.1
XP_016860439.1
5
551
XM_017004951.1
XP_016860440.1
5
506
XM_017004952.1
XP_016860441.1
5
950
XM_017004953.1
XP_016860442.1
5
522
XM_017004954.1
XP_016860443.1
6
610
XM_017004955.1
XP_016860444.1
8
958
XM_017004956.1
XP_016860445.1
3
506
XM_017004957.1
XP_016860446.1
5
552
GenBank mRNA
AB201269.1
-
2
290
AB597184.1
-
2
290
AK091684.1
-
3
382
AK291357.1
-
5
484
AY281366.1
-
5
569
BC017741.2
-
6
576
4390876
BC061699.1
-
5
476
4608518
BX647436.1
-
3
251
BX647773.1
-
7
707
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline