Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024686.4 | NP_078962.4 | 12 | 1702 | ||
XM_005271208.2 | XP_005271265.1 | 11 | 1513 | |||
XM_011542159.2 | XP_011540461.1 | 12 | 1892 | |||
XM_017002347.1 | XP_016857836.1 | 13 | 1722 | |||
XM_017002348.1 | XP_016857837.1 | 12 | 1892 | |||
XM_017002349.1 | XP_016857838.1 | 11 | 1477 | |||
GenBank mRNA | AK026686.1 | - | 5 | 468 | ||
AK094174.1 | - | 7 | 1304 | |||
AK126792.1 | - | 12 | 1599 | |||
AK128604.1 | - | 9 | 1681 | |||
AK225574.1 | - | 5 | 468 | |||
AY170843.1 | - | 11 | 1476 | |||
BC028350.2 | - | 5 | 638 | |||
BC032829.1 | - | 11 | 1444 | |||
BC048970.1 | - | 12 | 1644 | 5295193 | ||
BC060878.1 | - | 12 | 1684 | 5270163 | ||
BG718305.1 | - | 5 | 644 |