Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025105.2 | NP_001020276.1 | 9 | 1443 | ||
NM_001271741.1 | NP_001258670.1 | 8 | 1359 | |||
NM_001271742.1 | NP_001258671.1 | 8 | 882 | |||
NM_001892.5 | NP_001883.4 | 8 | 1359 | |||
GenBank mRNA | AF218004.1 | - | 8 | 1157 | ||
AK293761.1 | - | 8 | 849 | |||
AK294099.1 | - | 6 | 470 | |||
AK294942.1 | - | 9 | 946 | |||
AK303794.1 | - | 9 | 891 | |||
BC008717.2 | - | 8 | 901 | 2822250 | ||
BT019829.1 | - | 8 | 771 | |||
BT019830.1 | - | 8 | 771 | |||
CR542206.1 | - | 8 | 771 | |||
DQ082865.1 | - | 9 | 855 | |||
EU668329.1 | - | 8 | 771 | |||
L37042.1 | - | 8 | 982 | |||
X80693.1 | - | 8 | 896 |