Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024734.3 | NP_079010.2 | 11, 12 | 2893 | ||
XM_011537158.1 | XP_011535460.1 | 11, 12 | 2889 | |||
XM_017021646.1 | XP_016877135.1 | 11, 12 | 3059 | |||
GenBank mRNA | AB047979.1 | - | 11, 12 | 2850 | ||
AK023828.1 | - | 3, 4 | 1859 | |||
AK314315.1 | - | 11, 12 | 2891 | |||
BC014554.2 | - | 3, 4 | 1433 | |||
BC025251.1 | - | 3, 4 | 1832 | |||
BC068482.1 | - | 11, 12 | 2842 | 4375469 |