siRNA Details
Gene Symbol: VCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164097.1 | NP_001157569.1 | 7 | 6205 | ||
NM_004385.4 | NP_004376.2 | 8 | 9166 | |||
GenBank mRNA | AB209491.1 | - | 8 | 9049 | ||
J02814.1 | - | 1 | 686 | |||
U16306.1 | - | 8 | 9076 | |||
X15998.1 | - | 7 | 6115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
