Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171967.1 | NP_001165438.1 | 2, 3 | 348 | ||
NM_024805.2 | NP_079081.2 | 2, 3 | 348 | |||
GenBank mRNA | AF261073.2 | - | 2, 3 | 228 | ||
AF439745.2 | - | 2, 3 | 228 | |||
AK024825.1 | - | 2, 3 | 336 | |||
BC014195.2 | - | 2, 3 | 210 | 3938406 | ||
BC057772.1 | - | 2, 3 | 216 | 30337652 |