Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H14 SIRNA Assays ›
Gene Symbol:
ZC3H14
Gene Name:
zinc finger CCCH-type containing 14
Gene Aliases:
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location:
Chr.14: 88562909 - 88614720 on Build GRCh38
UniGene ID:
Hs.684046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36559
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001160103.1
NP_001153575.1
15
2278
NM_001160104.1
NP_001153576.1
15
2263
NM_001326295.1
NP_001313224.1
13
1888
NM_001326296.1
NP_001313225.1
14
2203
NM_001326297.1
NP_001313226.1
15
2462
NM_001326298.1
NP_001313227.1
12
1813
NM_001326299.1
NP_001313228.1
14
2188
NM_001326300.1
NP_001313229.1
14
2166
NM_001326301.1
NP_001313230.1
14
2387
NM_001326302.1
NP_001313231.1
14
2163
NM_001326303.1
NP_001313232.1
12
1991
NM_001326304.1
NP_001313233.1
12
1770
NM_001326305.1
NP_001313234.1
13
2091
NM_001326306.1
NP_001313235.1
11
1654
NM_001326307.1
NP_001313236.1
14
2206
NM_001326308.1
NP_001313237.1
12
1773
NM_001326309.1
NP_001313238.1
11
1698
NM_001326310.1
NP_001313239.1
15
2266
NM_001326311.1
NP_001313240.1
11
1695
NM_001326312.1
NP_001313241.1
14
2191
NM_001326313.1
NP_001313242.1
11
1657
NM_001326314.1
NP_001313243.1
13
2088
NM_001326315.1
NP_001313244.1
15
2459
NM_001326316.1
NP_001313245.1
15
2126
NM_024824.4
NP_079100.2
15
2281
NM_207660.3
NP_997543.1
12
1810
NM_207661.2
NP_997544.1
13
1736
NM_207662.3
NP_997545.2
5
1107
NR_136936.1
14
2121
XM_005268067.4
XP_005268124.1
15
2261
XM_005268068.4
XP_005268125.1
14
2201
XM_005268069.4
XP_005268126.1
13
1883
XM_005268070.4
XP_005268127.1
13
1880
XM_005268071.4
XP_005268128.1
12
1793
XM_005268073.3
XP_005268130.1
7
1503
XM_006720257.2
XP_006720320.1
6
1428
XM_011537160.2
XP_011535462.1
15
2389
XM_011537161.2
XP_011535463.1
14
2120
XM_017021648.1
XP_016877137.1
14
2198
XM_017021649.1
XP_016877138.1
14
2186
XM_017021650.1
XP_016877139.1
14
2183
GenBank mRNA
AF155107.1
-
4
369
AF474376.1
-
4
976
AK021868.1
-
12
1657
AK090809.1
-
1
1341
AK302136.1
-
12
1628
AK303123.1
-
14
1729
AK304275.1
-
15
2134
AL834215.1
-
13
1736
AY578060.1
-
15
2165
AY578061.1
-
12
1749
AY578062.1
-
13
1736
AY578063.1
-
7
814
BC011793.1
-
15
2147
4298961
BC023641.2
-
11
1784
BC027607.2
-
5
1093
BX248265.1
-
11
1800
BX248281.1
-
7
882
BX248769.1
-
2
171
BX640716.1
-
15
2160
BX640876.1
-
4
474
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: DQ303482.This siRNA also targets the following transcripts in Mouse: NM_001008506, NM_001160107, NM_001160108, NM_029334, BC024856, and others.
Gene Ontology Categories:
Function(s)
Process(es)
regulation of mRNA stability
negative regulation of mRNA polyadenylation
protein binding
poly(A) binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-jgtjs:80/10.239.162.121:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0