Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THNSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024838.4 | NP_079114.3 | 3 | 1536 | ||
XM_005252597.2 | XP_005252654.1 | 3 | 1612 | |||
XM_017016665.1 | XP_016872154.1 | 4 | 1622 | |||
GenBank mRNA | AK094362.1 | - | 1 | 755 | ||
AK125249.1 | - | 3 | 1464 | |||
BC016697.1 | - | 1 | 26 | |||
BC035132.2 | - | 3 | 1457 | 5263853 | ||
BX647989.1 | - | 3 | 1449 |