Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308235.1 | NP_001295164.1 | 6 | 1894 | ||
NM_001308236.1 | NP_001295165.1 | 6 | 1717 | |||
NM_024872.2 | NP_079148.2 | 6 | 1673 | |||
XM_005265985.3 | XP_005266042.1 | 6 | 1730 | |||
XM_011534654.2 | XP_011532956.1 | 6 | 1607 | |||
XM_011534655.2 | XP_011532957.1 | 6 | 1783 | |||
XM_011534656.2 | XP_011532958.1 | 6 | 1584 | |||
XM_011534657.2 | XP_011532959.1 | 5 | 1434 | |||
XM_017009883.1 | XP_016865372.1 | 4 | 1405 | |||
GenBank mRNA | AK026223.1 | - | 6 | 1668 |