Search
Search

ZNF442| Gene Symbol: ZNF442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030824.2 | NP_110451.1 | 6 | 1495 | ||
| XM_006722908.3 | XP_006722971.1 | 5 | 975 | |||
| XM_017027316.1 | XP_016882805.1 | 4 | 923 | |||
| XM_017027317.1 | XP_016882806.1 | 4 | 825 | |||
| XM_017027318.1 | XP_016882807.1 | 4 | 1428 | |||
| GenBank mRNA | AK024418.1 | - | 6 | 1467 | ||
| AK295923.1 | - | 3 | 788 | |||