Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRMT2B SIRNA Assays ›
Gene Symbol:
TRMT2B
Gene Name:
tRNA methyltransferase 2 homolog B
Gene Aliases:
CXorf34, dJ341D10.3
Chromosome Location:
Chr.X: 100973358 - 101052118 on Build GRCh38
UniGene ID:
Hs.496501
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36759
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRMT2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001167970.1
NP_001161442.1
10
1379
NM_001167971.1
NP_001161443.1
10
1360
NM_001167972.1
NP_001161444.1
9
1322
NM_024917.5
NP_079193.2
10
1772
XM_005262195.2
XP_005262252.1
11
1495
XM_005262196.2
XP_005262253.1
9
1290
XM_005262201.2
XP_005262258.1
7
870
XM_006724705.2
XP_006724768.1
10
1434
XM_011531042.2
XP_011529344.1
7
988
XM_011531044.2
XP_011529346.1
9
1049
XM_017029858.1
XP_016885347.1
10
1767
XM_017029859.1
XP_016885348.1
9
1082
XM_017029860.1
XP_016885349.1
10
1381
XM_017029861.1
XP_016885350.1
9
1291
XM_017029862.1
XP_016885351.1
9
1632
XM_017029863.1
XP_016885352.1
11
1304
XM_017029864.1
XP_016885353.1
9
1496
XM_017029865.1
XP_016885354.1
9
1124
XM_017029866.1
XP_016885355.1
10
1187
XM_017029867.1
XP_016885356.1
10
1224
XM_017029868.1
XP_016885357.1
9
1097
XM_017029869.1
XP_016885358.1
8
1441
XM_017029870.1
XP_016885359.1
8
1031
XM_017029871.1
XP_016885360.1
8
1013
XM_017029872.1
XP_016885361.1
9
1124
XM_017029873.1
XP_016885362.1
9
1099
XM_017029874.1
XP_016885363.1
8
1441
XM_017029875.1
XP_016885364.1
7
988
GenBank mRNA
AK022749.1
-
9
1135
AK315780.1
-
10
1529
AL832849.1
-
10
1298
BC007526.1
-
9
1278
2988022
BC008067.2
-
3
321
BC009437.1
-
9
1265
3504468
BC020116.1
-
10
1333
4342798
BC034272.2
-
10
1737
4933351
BX647969.1
-
9
1319
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
RNA methylation
tRNA processing
protein binding
S-adenosylmethionine-dependent tRNA (m5U54) methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0