Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025004.2 | NP_079280.2 | 13 | 2583 | ||
XM_017018346.1 | XP_016873835.1 | 14 | 2703 | |||
XM_017018347.1 | XP_016873836.1 | 15 | 2613 | |||
XM_017018348.1 | XP_016873837.1 | 13 | 2595 | |||
XM_017018349.1 | XP_016873838.1 | 13 | 3844 | |||
XM_017018350.1 | XP_016873839.1 | 12 | 2460 | |||
XM_017018351.1 | XP_016873840.1 | 11 | 2352 | |||
XM_017018352.1 | XP_016873841.1 | 13 | 2353 | |||
XM_017018353.1 | XP_016873842.1 | 10 | 2277 | |||
GenBank mRNA | AK023277.1 | - | 13 | 2583 | ||
BC018540.1 | - | 13 | 2497 | |||
BU569785.1 | - | 2 | 145 | |||
DB516555.2 | - | 2 | 36 |