Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025041.2 | NP_079317.2 | 1 | 1636 | ||
GenBank mRNA | AK025826.1 | - | 1 | 1498 | ||
AK225383.1 | - | 1 | 1498 | |||
BC051274.1 | - | 1 | 1636 | |||
BC064502.1 | - | 1 | 1636 | |||
BC082257.1 | - | 1 | 1636 |