Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTBP2 SIRNA Assays ›
Gene Symbol:
CTBP2
Gene Name:
C-terminal binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 124987849 - 125161061 on Build GRCh38
UniGene ID:
Hs.501345
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s3702
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083914.2
NP_001077383.1
5, 6
806
NM_001290214.2
NP_001277143.1
5, 6
791
NM_001290215.2
NP_001277144.1
5, 6
1055
NM_001321012.1
NP_001307941.1
5, 6
750
NM_001321013.1
NP_001307942.1
4, 5
951
NM_001321014.1
NP_001307943.1
5, 6
1052
NM_001329.3
NP_001320.1
5, 6
794
NM_022802.2
NP_073713.2
3, 4
2111
XM_005269561.2
XP_005269618.1
6, 7
977
XM_005269564.2
XP_005269621.1
6, 7
1185
XM_005269567.2
XP_005269624.1
6, 7
1184
XM_005269568.4
XP_005269625.1
6, 7
912
XM_005269569.2
XP_005269626.1
5, 6
873
XM_005269571.2
XP_005269628.1
4, 5
632
XM_005269572.3
XP_005269629.1
4, 5
680
XM_006717642.2
XP_006717705.1
7, 8
1106
XM_011539349.2
XP_011537651.1
3, 4
888
XM_011539351.1
XP_011537653.1
8, 9
1207
XM_011539353.1
XP_011537655.1
7, 8
1078
XM_011539354.1
XP_011537656.1
6, 7
1002
XM_011539355.1
XP_011537657.1
5, 6
1080
XM_011539357.1
XP_011537659.1
5, 6
818
XM_011539358.1
XP_011537660.1
4, 5
896
XM_017015756.1
XP_016871245.1
6, 7
848
XM_017015757.1
XP_016871246.1
5, 6
2574
GenBank mRNA
AF016507.1
-
5, 6
711
AF222711.1
-
3, 4
2128
AK290390.1
-
5, 6
655
AL833398.1
-
5, 6
766
BC002486.2
-
5, 6
767
BC037900.2
-
3, 4
775
5276086
BC047018.1
-
5, 6
771
BC052276.1
-
5, 6
748
BC072020.1
-
5, 6
728
BM800106.1
-
3, 4
466
BT007012.1
-
3, 4
366
CV029885.1
-
3, 4
366
CX165167.1
-
5, 6
620
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of cell proliferation
viral genome replication
positive regulation of chromatin binding
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of retinoic acid receptor signaling pathway
white fat cell differentiation
oxidation-reduction process
chromatin binding
transcription coactivator activity
transcription corepressor activity
protein binding
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
protein homodimerization activity
retinoic acid receptor binding
NAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-cdcg2:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0