Search
Search

RNF34| Gene Symbol: RNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025126.3 | NP_079402.2 | 3 | 536 | ||
| NM_194271.2 | NP_919247.1 | 4 | 567 | |||
| GenBank mRNA | AB084914.1 | - | 2 | 409 | ||
| AF306709.1 | - | 3 | 578 | |||
| AK025439.1 | - | 3 | 501 | |||
| AK225285.1 | - | 3 | 501 | |||
| AK304366.1 | - | 4 | 556 | |||
| AL557811.3 | - | 4 | 577 | |||
| AY098934.1 | - | 2 | 406 | |||
| BC007826.2 | - | 3 | 461 | 3347885 | ||
| BT007283.1 | - | 2 | 406 | |||
| CR457342.1 | - | 2 | 406 | |||
| DB242808.1 | - | 3 | 537 | |||