Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTGF SIRNA Assays ›
          Gene Symbol:
          CTGF
          Gene Name:
          connective tissue growth factor
          Gene Aliases:
          CCN2, HCS24, IGFBP8, NOV2
          Chromosome Location:
          Chr.6: 131948176 - 131951378 on Build GRCh38
          UniGene ID:
          Hs.410037
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s3707

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTGF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001901.2 NP_001892.1 5 979
          GenBank mRNA AK092280.1 - 5 830
          AK125220.1 - 5 761
          AK290884.1 - 5 973
          AK315207.1 - 5 973
          AY395801.1 - 5 896
          AY550024.1 - 5 886
          BC087839.1 - 5 955 6181580
          BT019794.1 - 5 773
          BT019795.1 - 5 773
          CR541734.1 - 5 773
          CR541759.1 - 5 773
          M92934.1 - 5 902
          U14750.1 - 4 553
          X78947.1 - 5 918

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          ossification
          angiogenesis
          regulation of cell growth
          tissue homeostasis
          transcription initiation from RNA polymerase II promoter
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          cell-cell signaling
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          epidermis development
          response to wounding
          response to glucose
          organ senescence
          positive regulation of gene expression
          negative regulation of gene expression
          cell migration
          cell differentiation
          lung development
          regulation of chondrocyte differentiation
          response to estradiol
          positive regulation of collagen biosynthetic process
          response to anoxia
          intracellular signal transduction
          chondrocyte proliferation
          response to amino acid
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to peptide hormone
          positive regulation of cell differentiation
          positive regulation of JNK cascade
          positive regulation of cell activation
          response to mineralocorticoid
          positive regulation of stress fiber assembly
          cytosolic calcium ion transport
          positive regulation of cardiac muscle contraction
          negative regulation of cell death
          extracellular matrix constituent secretion
          positive regulation of G0 to G1 transition
          positive regulation of ERK1 and ERK2 cascade
          response to fatty acid
          DNA biosynthetic process
          reactive oxygen species metabolic process
          fibronectin binding
          integrin binding
          protein binding
          insulin-like growth factor binding
          protein C-terminus binding
          growth factor activity
          heparin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0