Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025194.2 | NP_079470.1 | 3 | 1316 | ||
XM_017027324.1 | XP_016882813.1 | 4 | 500 | |||
GenBank mRNA | AJ290975.1 | - | 3 | 1283 | ||
AK024596.1 | - | 3 | 351 | |||
BC060788.1 | - | 3 | 1316 | 30332196 | ||
D38169.1 | - | 3 | 1047 |