Search
Search

CTSS| Gene Symbol: CTSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199739.1 | NP_001186668.1 | 5 | 787 | ||
| NM_004079.4 | NP_004070.3 | 6 | 937 | |||
| GenBank mRNA | AK226095.1 | - | 6 | 799 | ||
| AK301472.1 | - | 5 | 622 | |||
| AK314482.1 | - | 6 | 778 | |||
| BC002642.2 | - | 6 | 738 | 3610589 | ||
| CR541676.1 | - | 5 | 676 | |||
| M86553.1 | - | 5 | 682 | |||
| M90696.1 | - | 6 | 812 | |||
| S93414.1 | - | 5 | 682 | |||