Search
Search

CTSS| Gene Symbol: CTSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199739.1 | NP_001186668.1 | 4 | 688 | ||
| NM_004079.4 | NP_004070.3 | 5 | 838 | |||
| GenBank mRNA | AK226095.1 | - | 5 | 700 | ||
| AK301472.1 | - | 4 | 523 | |||
| AK314482.1 | - | 5 | 679 | |||
| BC002642.2 | - | 5 | 639 | 3610589 | ||
| CR541676.1 | - | 4 | 577 | |||
| M86553.1 | - | 4 | 583 | |||
| M90696.1 | - | 5 | 713 | |||
| S93414.1 | - | 4 | 583 | |||