Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040177.2 | NP_001035267.1 | 6 | 764 | ||
NR_073125.1 | 6 | 764 | ||||
NR_073126.1 | 6 | 678 | ||||
XM_011519715.2 | XP_011518017.1 | 6 | 1061 | |||
XM_011519718.2 | XP_011518020.1 | 6 | 1061 | |||
XM_011519719.2 | XP_011518021.1 | 6 | 1061 | |||
XM_011519720.2 | XP_011518022.1 | 6 | 1060 | |||
XM_011519722.2 | XP_011518024.1 | 6 | 1061 | |||
XM_011519725.2 | XP_011518027.1 | 6 | 1058 | |||
XM_017016743.1 | XP_016872232.1 | 6 | 1062 | |||
XM_017016744.1 | XP_016872233.1 | 6 | 1061 | |||
XM_017016745.1 | XP_016872234.1 | 6 | 847 | |||
GenBank mRNA | AB055603.1 | - | 6 | 616 | ||
AF263242.1 | - | 6 | 636 | |||
AK074211.1 | - | 6 | 678 | |||
BC002862.2 | - | 6 | 707 | 3941289 |