Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317238.1 | NP_001304167.1 | 11 | 1525 | ||
NM_031921.5 | NP_114127.3 | 13 | 1408 | |||
XM_005244806.3 | XP_005244863.1 | 13 | 1431 | |||
XM_011542241.2 | XP_011540543.1 | 13 | 1431 | |||
XM_011542243.2 | XP_011540545.1 | 12 | 1144 | |||
XM_011542244.1 | XP_011540546.1 | 13 | 1575 | |||
GenBank mRNA | AB033099.1 | - | 11 | 1614 | ||
AF343078.1 | - | 13 | 1341 | |||
AK024028.1 | - | 11 | 993 | |||
AK127857.1 | - | 6 | 2059 | |||
AK290586.1 | - | 13 | 1400 | |||
AL834179.1 | - | 4 | 1194 | |||
BC002542.2 | - | 13 | 1334 | 3138578 | ||
BC018701.1 | - | 13 | 1334 | 3139886 |