Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031935.2 | NP_114141.2 | 29 | 4592 | ||
XM_011510038.2 | XP_011508340.1 | 29 | 4609 | |||
XM_011510041.2 | XP_011508343.1 | 29 | 4617 | |||
XM_017002437.1 | XP_016857926.1 | 18 | 2542 | |||
GenBank mRNA | AF156100.1 | - | 29 | 4592 | ||
AK056557.1 | - | 12 | 1691 |