Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024380.1 | NP_001019551.1 | 4 | 729 | ||
NM_001024381.1 | NP_001019552.1 | 4 | 628 | |||
NM_031940.3 | NP_114146.3 | 4 | 935 | |||
NM_078473.2 | NP_510882.1 | 4 | 546 | |||
XM_005273657.3 | XP_005273714.1 | 4 | 829 | |||
XM_006716408.3 | XP_006716471.1 | 4 | 711 | |||
XM_011544667.2 | XP_011542969.1 | 4 | 1071 | |||
GenBank mRNA | AF353991.1 | - | 4 | 516 | ||
AK075027.1 | - | 4 | 907 | |||
AK075155.1 | - | 4 | 930 | |||
AK314703.1 | - | 4 | 535 | |||
AL834224.1 | - | 4 | 589 | |||
BC004878.2 | - | 4 | 898 | 3688720 | ||
BC109049.1 | - | 4 | 514 | 40030485 | ||
BC109050.1 | - | 4 | 512 | 40030488 | ||
BG424960.1 | - | 546 |