Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000771.3 | NP_000762.2 | 5 | 790 | ||
XM_017015758.1 | XP_016871247.1 | 6 | 1210 | |||
GenBank mRNA | AK289420.1 | - | 5 | 790 | ||
AK298458.1 | - | 5 | 790 | |||
BC125054.1 | - | 5 | 765 | 40116637 | ||
D00173.1 | - | 5 | 756 | |||
M15331.1 | - | 5 | 750 | |||
M21939.1 | - | 5 | 751 | |||
M21940.1 | - | 3 | 444 | |||
M61855.1 | - | 5 | 777 | |||
M61857.1 | - | 5 | 775 | |||
S46963.1 | - | 5 | 728 |