Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303096.1 | NP_001290025.1 | 20 | 2220 | ||
NM_032168.2 | NP_115544.1 | 19 | 2186 | |||
GenBank mRNA | AK022581.1 | - | 19 | 2133 | ||
AK022607.1 | - | 19 | 2145 | |||
AK023276.1 | - | 16 | 1825 | |||
AK091546.1 | - | 20 | 2147 | |||
AK290445.1 | - | 19 | 2137 | |||
AL122097.1 | - | 15 | 1753 | |||
BC006816.1 | - | 4 | 380 | |||
BC018689.1 | - | 2 | 345 | |||
BC040567.1 | - | 19 | 2186 | 5267120 | ||
CR749440.1 | - | 19 | 3795 |