Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM175A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139076.2 | NP_620775.2 | 8 | 788 | ||
XM_005263280.4 | XP_005263337.1 | 9 | 921 | |||
XM_017008694.1 | XP_016864183.1 | 7 | 694 | |||
GenBank mRNA | AK021582.1 | - | 3 | 996 | ||
AK022704.1 | - | 8 | 774 | |||
AK023676.1 | - | 7 | 688 | |||
AY358576.1 | - | 8 | 739 | |||
BC039573.1 | - | 8 | 735 | 6045433 | ||
EF531340.1 | - | 8 | 723 |