siRNA Details
Gene Symbol: POLR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137604.2 | NP_001131076.1 | 4 | 1128 | ||
NM_001282772.1 | NP_001269701.1 | 6 | 946 | |||
NM_001282774.1 | NP_001269703.1 | 5 | 1296 | |||
NM_001282776.1 | NP_001269705.1 | 2 | 848 | |||
NM_001282777.1 | NP_001269706.1 | 6 | 1388 | |||
NM_001282779.1 | NP_001269708.1 | 5 | 1225 | |||
NM_019014.5 | NP_061887.2 | 5 | 1296 | |||
GenBank mRNA | AK001678.1 | - | 5 | 768 | ||
AK022533.1 | - | 4 | 578 | |||
AK128044.1 | - | 5 | 1296 | |||
AK294027.1 | - | 5 | 697 | |||
AK301178.1 | - | 2 | 320 | |||
AK302814.1 | - | 6 | 946 | |||
BC110833.1 | - | 5 | 985 | 6053210 | ||
BX647360.1 | - | 6 | 1114 | |||
BX647683.1 | - | 5 | 675 |

More Information
Panther Classification:
Molecular Function -





