Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SGIP1 SIRNA Assays ›
          Gene Symbol:
          SGIP1
          Gene Name:
          SH3 domain GRB2 like endophilin interacting protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 66533272 - 66751139 on Build GRCh38
          UniGene ID:
          Hs.132121
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s38727

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SGIP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001308203.1 NP_001295132.1 16, 17 1370
          NM_032291.3 NP_115667.2 19, 20 2147
          XM_005271264.3 XP_005271321.1 19, 20 1981
          XM_005271268.3 XP_005271325.1 17, 18 1396
          XM_005271270.4 XP_005271327.1 16, 17 1703
          XM_006710961.2 XP_006711024.1 19, 20 1987
          XM_006710966.2 XP_006711029.1 18, 19 1921
          XM_006710967.2 XP_006711030.1 18, 19 1909
          XM_006710969.2 XP_006711032.1 18, 19 1897
          XM_006710971.2 XP_006711034.1 18, 19 1486
          XM_006710972.2 XP_006711035.1 18, 19 1480
          XM_006710973.2 XP_006711036.1 16, 17 1462
          XM_006710974.2 XP_006711037.1 15, 16 1378
          XM_011542291.1 XP_011540593.1 18, 19 1643
          XM_011542292.1 XP_011540594.1 17, 18 1583
          XM_011542293.1 XP_011540595.1 14, 15 1312
          XM_017002505.1 XP_016857994.1 20, 21 2035
          XM_017002506.1 XP_016857995.1 21, 22 2158
          XM_017002507.1 XP_016857996.1 20, 21 2210
          XM_017002508.1 XP_016857997.1 21, 22 2156
          XM_017002509.1 XP_016857998.1 18, 19 2011
          XM_017002510.1 XP_016857999.1 19, 20 2134
          XM_017002511.1 XP_016858000.1 20, 21 2092
          XM_017002512.1 XP_016858001.1 20, 21 1965
          XM_017002513.1 XP_016858002.1 19, 20 2144
          XM_017002514.1 XP_016858003.1 19, 20 1963
          XM_017002515.1 XP_016858004.1 17, 18 1939
          XM_017002516.1 XP_016858005.1 18, 19 2062
          XM_017002517.1 XP_016858006.1 19, 20 2031
          XM_017002518.1 XP_016858007.1 19, 20 2019
          XM_017002519.1 XP_016858008.1 19, 20 2044
          XM_017002520.1 XP_016858009.1 17, 18 1885
          XM_017002521.1 XP_016858010.1 17, 18 1837
          XM_017002522.1 XP_016858011.1 18, 19 1959
          XM_017002523.1 XP_016858012.1 18, 19 1983
          XM_017002524.1 XP_016858013.1 18, 19 1626
          XM_017002525.1 XP_016858014.1 17, 18 1565
          XM_017002526.1 XP_016858015.1 16, 17 1444
          XM_017002527.1 XP_016858016.1 17, 18 1468
          XM_017002528.1 XP_016858017.1 17, 18 1554
          XM_017002529.1 XP_016858018.1 17, 18 1420
          XM_017002530.1 XP_016858019.1 16, 17 1396
          XM_017002531.1 XP_016858020.1 15, 16 1396
          XM_017002532.1 XP_016858021.1 16, 17 1390
          XM_017002533.1 XP_016858022.1 16, 17 1498
          XM_017002534.1 XP_016858023.1 15, 16 1372
          XM_017002535.1 XP_016858024.1 16, 17 1486
          XM_017002536.1 XP_016858025.1 16, 17 1336
          XM_017002537.1 XP_016858026.1 15, 16 1425
          GenBank mRNA AB210039.1 - 19, 20 1929
          AK090697.1 - 10, 11 927
          AK125044.1 - 2, 3 914
          AK298970.1 - 16, 17 1333
          AK299941.1 - 17, 18 1407
          AL136561.1 - 19, 20 1821
          BC040516.1 - 21, 22 1975 5275011
          BX640813.1 - 16, 17 1347
          CR749541.1 - 16, 17 1558

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          positive regulation of receptor-mediated endocytosis
          membrane tubulation
          protein binding
          phospholipid binding
          microtubule binding
          SH3 domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0